Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Amino-2-Naphthol-4-Sulfonic Acid Solution, For Silica, Certified, LabChem™

Catalog No. LC108907
Change view
Click to view available options
Quantity:
125 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC108907 125 mL
LC108901 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LC108907 Supplier LabChem, Inc. Supplier No. LC108907
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7732-18-5 , 7631-90-5 , 7757-83-7 , 116-63-2
Synonym water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide

Specifications

Color Brown
CAS Max % 84.25, 15, 0.5, 0.25
Linear Formula H2N(HO)C10H5SO3H
Solubility Information Soluble in water
Formula Weight 239.26
Grade Certified
Identification Passes Test
Packaging White Poly
Suitability Passes Test
Decomposition Information Sulfur compounds; Carbon monoxide; Carbon dioxide
Quantity 125 mL
Physical Form Liquid
Chemical Name or Material 1-Amino-2-Naphthol-4-Sulfonic Acid Solution, For Silica
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Recommended Storage Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.