Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Benzofuran-2-sulfonyl chloride, Thermo Scientific™

Catalog No. CC06603DA
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC06603DA 1 g Amber Glass Bottle
CC06603CB 250 mg Amber Glass Bottle
CC06603DE 5 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC06603DA Supplier Thermo Scientific Chemicals Supplier No. CC06603DA
Add to Cart

Chemical Identifiers

CAS 17070-58-5
Molecular Formula C8H5ClO3S
Molecular Weight (g/mol) 216.635
InChI Key RWCKBBSBRTUUHR-UHFFFAOYSA-N
Synonym benzofuran-2-sulfonyl chloride, 2-benzofuransulfonylchloride, 2-benzofuransulfonyl chloride, benzo b furan-2-sulfonyl chloride, pubchem7024, benzo d furan-2-ylchlorosulfone
PubChem CID 2795172
IUPAC Name 1-benzofuran-2-sulfonyl chloride
SMILES C1=CC=C2C(=C1)C=C(O2)S(=O)(=O)Cl

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 216.64
Percent Purity 97%
Chemical Name or Material 1-Benzofuran-2-sulfonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.