Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Benzothiophene-5-carboxylic acid, 97%, Thermo Scientific™

Catalog No. CC24801DE
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC24801DE 5 g Amber Glass Bottle
CC24801CB 250 mg Amber Glass Bottle
CC24801DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC24801DE Supplier Thermo Scientific Chemicals Supplier No. CC24801DE
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2060-64-2
Molecular Formula C9H6O2S
Molecular Weight (g/mol) 178.21
MDL Number MFCD04974042
InChI Key SNBYTKLWZRHESA-UHFFFAOYSA-N
Synonym benzo b thiophene-5-carboxylic acid, buttpark 98\04-76, pubchem20599, 5-carboxybenzo b thiophene, benzothiophene-5-carboxylic acid, benzo b thiophene-5-carboxylicacid, 1-benzo b thiophene-5-carboxylic acid
PubChem CID 2795444
IUPAC Name 1-benzothiophene-5-carboxylic acid
SMILES OC(=O)C1=CC=C2SC=CC2=C1

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 178.21
Percent Purity 95%
Chemical Name or Material 1-Benzothiophene-5-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.