Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Benzyl-3-acetamidopyrrolidine 98.0+%, TCI America™
SDP

Catalog No. B135525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B135525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B135525G Supplier TCI America Supplier No. B135525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 28506-01-6
Molecular Formula C13H18N2O
Molecular Weight (g/mol) 218.3
MDL Number MFCD00059037
InChI Key CMSWETNAAPYFSH-UHFFFAOYSA-N
Synonym 3-Acetamido-1-benzylpyrrolidine
PubChem CID 4198339
IUPAC Name N-(1-benzylpyrrolidin-3-yl)acetamide
SMILES CC(=O)NC1CCN(C1)CC2=CC=CC=C2

Specifications

Melting Point 91°C
Color White-Yellow
Quantity 25 g
Formula Weight 218.30
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Benzyl-3-acetamidopyrrolidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.