Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Benzyl-3-methyl-4-piperidone 98.0+%, TCI America™
SDP

Catalog No. B44651G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B44651G 1 g
B44655G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B44651G Supplier TCI America Supplier No. B44651G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 34737-89-8
Molecular Formula C13H17NO
Molecular Weight (g/mol) 203.285
MDL Number MFCD00044806
InChI Key OVQAJYCAXPHYNV-UHFFFAOYSA-N
Synonym 1-benzyl-3-methyl-4-piperidone, 4-piperidinone, 3-methyl-1-phenylmethyl, 1-benzyl-3-methyl-4-piperidinone, 4-piperidone, 1-benzyl-3-methyl, n-benzyl-3-methyl-4-piperidone, 3-methyl-1-phenylmethyl-4-piperidinone, 3-methyl-1-benzylpiperidin-4-one, ksc506m4r, 1-benzyl-3-methyl-piperidin-4one, 1-benzyl-3-methyl-piperidin4-one
PubChem CID 560965
IUPAC Name 1-benzyl-3-methylpiperidin-4-one
SMILES CC1CN(CCC1=O)CC2=CC=CC=C2

Specifications

Color Yellow
Boiling Point 99°C
Quantity 1 g
UN Number 1760
Formula Weight 203.29
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 1-Benzyl-3-methyl-4-piperidone
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.