Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Benzyl-3-(tert-butoxycarbonylamino)pyrrolidine 98.0+%, TCI America™
SDP

Catalog No. B135710G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B135710G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B135710G Supplier TCI America Supplier No. B135710G

Chemical Identifiers

CAS 99735-30-5
Molecular Formula C16H24N2O2
Molecular Weight (g/mol) 276.38
MDL Number MFCD00059039
InChI Key PHOIDJGLYWEUEK-UHFFFAOYNA-N
PubChem CID 3575823
IUPAC Name tert-butyl N-(1-benzylpyrrolidin-3-yl)carbamate
SMILES CC(C)(C)OC(=O)NC1CCN(CC2=CC=CC=C2)C1

Specifications

Melting Point 113°C
Color White-Yellow
Quantity 10 g
Formula Weight 276.38
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Benzyl-3-(tert-butoxycarbonylamino)pyrrolidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.