Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Benzyl-4-piperidone 98.0+%, TCI America™
SDP

Catalog No. B1027500ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B1027500ML 500 mL
B102725ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B1027500ML Supplier TCI America Supplier No. B1027500ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3612-20-2
Molecular Formula C12H15NO
Molecular Weight (g/mol) 189.258
MDL Number MFCD00006192
InChI Key SJZKULRDWHPHGG-UHFFFAOYSA-N
Synonym 1-benzyl-4-piperidone, n-benzyl-4-piperidone, 4-piperidinone, 1-phenylmethyl, 1-benzyl-4-piperidinone, 1-benzylpiperidone, 1-benzyl-4-oxopiperidine, 1-benzyl-piperidin-4-one, n-benzyl-4-piperidine, n-benzyl-4-piperidinone, n-benzylpiperidin-4-one
PubChem CID 19220
IUPAC Name 1-benzylpiperidin-4-one
SMILES C1CN(CCC1=O)CC2=CC=CC=C2

Specifications

Color Yellow
Boiling Point 158°C
Quantity 500 mL
Formula Weight 189.26
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 1-Benzyl-4-piperidone
EINECSNumber (5)-3734
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.