Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Benzyl N-Carbobenzoxy-L-glutamate 98.0+%, TCI America™
SDP

Catalog No. B398925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B398925G 25 g
B39895G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B398925G Supplier TCI America Supplier No. B398925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3705-42-8
Molecular Formula C20H21NO6
Molecular Weight (g/mol) 371.389
MDL Number MFCD00077013
InChI Key VWHKODOUMSMUAF-KRWDZBQOSA-N
Synonym z-glu-obzl, cbz-glu-obzl, cbz-l-glutamic acid 1-benzyl ester, z-l-glu-obzl, n-cbz-l-glutamic acid benzyl ester, n-carbobenzyloxy-l-glutamic acid o-benzyl ester, s-5-benzyloxy-4-benzyloxy carbonyl amino-5-oxopentanoic acid, 4s-5-benzyloxy-4-benzyloxy carbonyl amino-5-oxopentanoic acid, n-benzyloxycarbonyl-l-glutamic acid, a-benzyl ester, z-l-glutamic acid alpha-benzyl ester
PubChem CID 6994031
IUPAC Name (4S)-5-oxo-5-phenylmethoxy-4-(phenylmethoxycarbonylamino)pentanoic acid
SMILES C1=CC=C(C=C1)COC(=O)C(CCC(=O)O)NC(=O)OCC2=CC=CC=C2

Specifications

Melting Point 100°C
Color White
Quantity 25 g
Formula Weight 371.39
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Benzyl N-Carbobenzoxy-L-glutamate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.