Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Benzyl N-(tert-Butoxycarbonyl)-D-glutamate 98.0+%, TCI America™
SDP

Catalog No. B430325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B430325G 25 g
B43035G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B430325G Supplier TCI America Supplier No. B430325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 34404-30-3
Molecular Formula C17H23NO6
Molecular Weight (g/mol) 337.372
MDL Number MFCD00038266
InChI Key CVZUKWBYQQYBTF-CYBMUJFWSA-N
Synonym boc-d-glu-obzl, boc-d-glutamic acid 1-benzyl ester, n-boc-d-glutamic acid 1-benzyl ester, n-boc-d-glutamic acid benzyl ester, 1-benzyl n-boc-l-glutamate, n-tert-butoxycarbonyl-d-glutamic acid 1-benzyl ester, 4r-5-benzyloxy-4-tert-butoxycarbonyl amino-5-oxopentanoic acid, boc-d-glu-obn, pubchem12942, 1-benzyl n-boc-d-glutamate
PubChem CID 7010515
IUPAC Name (4R)-4-[(2-methylpropan-2-yl)oxycarbonylamino]-5-oxo-5-phenylmethoxypentanoic acid
SMILES CC(C)(C)OC(=O)NC(CCC(=O)O)C(=O)OCC1=CC=CC=C1

Specifications

Melting Point 98°C
Color White
Quantity 25 g
Formula Weight 337.37
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Benzyl N-(tert-Butoxycarbonyl)-D-glutamate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.