Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(Benzyloxycarbonyl)pyrrolidine-3-carboxylic acid, 97%, Thermo Scientific™

Catalog No. AAH6472806
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6472806 5 g
AAH64728MD 250 mg
AAH6472803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6472806 Supplier Thermo Scientific Chemicals Supplier No. H6472806

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 188527-21-1
Molecular Formula C13H15NO4
Molecular Weight (g/mol) 249.266
MDL Number MFCD02179036
InChI Key JSASVUTVTRNJHA-UHFFFAOYSA-N
Synonym 1-n-cbz-pyrrolidine-3-carboxylic acid, n-cbz-pyrrolidine-3-carboxylic acid, 1-benzyloxycarbonyl pyrrolidine-3-carboxylic acid, 1-benzyloxy carbonyl pyrrolidine-3-carboxylic acid, 1-cbz-pyrrolidine-3-carboxylic acid, 1-n-cbz-beta-proline, 1-n-cbz-belta-proline, 1-cbz-3-pyrrolidinecarboxylic acid, 3-carboxy-1-cbz-pyrrolidine
PubChem CID 18538483
IUPAC Name 1-phenylmethoxycarbonylpyrrolidine-3-carboxylic acid
SMILES C1CN(CC1C(=O)O)C(=O)OCC2=CC=CC=C2

Specifications

Quantity 5 g
Beilstein 13268300
Formula Weight 249.27
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 1-(Benzyloxycarbonyl)pyrrolidine-3-carboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.