Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Benzylpiperidine 98.0+%, TCI America™
SDP

Catalog No. B469825ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B469825ML 25 mL
B46985ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B469825ML Supplier TCI America Supplier No. B469825ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2905-56-8
Molecular Formula C12H18ClN
Molecular Weight (g/mol) 211.73
MDL Number MFCD00224901
InChI Key MPPIBJJDFLONMO-UHFFFAOYSA-N
Synonym piperidine, 1-phenylmethyl, benzylpiperidine, unii-7hze16210b, n-benzylpiperidine, benzyl-piperidine, n-benzyl piperidine, n-benzyl-piperidine, 1-benzyl-piperidine, acmc-20a2pl
PubChem CID 76190
IUPAC Name hydrogen 1-benzylpiperidine chloride
SMILES [H+].[Cl-].C(N1CCCCC1)C1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 66°C
Quantity 25 mL
Formula Weight 175.28
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 1-Benzylpiperidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.