Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Boc-2-piperidone, 99%

Catalog No. AAL1889603
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1889603 1 g
AAL1889606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1889603 Supplier Thermo Scientific Chemicals Supplier No. L1889603
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 85908-96-9 | C10H17NO3 | 199.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85908-96-9
Molecular Formula C10H17NO3
Molecular Weight (g/mol) 199.25
MDL Number MFCD02179046
InChI Key ULMHMJAEGZPQRY-UHFFFAOYSA-N
Synonym 1-boc-2-piperidone, n-boc-2-piperidone, 1-boc-1-azacyclohexan-2-one, 1-tert-butoxycarbonyl-2-piperidone, n-tert-butoxycarbonyl-2-piperidone, tert-butyl 2-oxo-1-piperidinecarboxylate, 1-piperidinecarboxylic acid, 2-oxo-, 1,1-dimethylethyl ester, tert-butyl 2-oxopiperidinecarboxylate, boc-piperidinone, n-boc piperidone
PubChem CID 7577838
ChEBI CHEBI:59556
IUPAC Name tert-butyl 2-oxopiperidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCCCC1=O

Specifications

Melting Point 34°C to 37°C
Quantity 1 g
Beilstein 4674955
Formula Weight 199.25
Percent Purity 99%
Chemical Name or Material 1-Boc-2-piperidone
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.