Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Boc-3-iodoazetidine, 97%

Catalog No. AAH5255806
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5255806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5255806 Supplier Thermo Scientific Chemicals Supplier No. H5255806
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 254454-54-1 | C8H14INO2 | 283.109 g/mol

1-Boc-3-iodoazetidine is used as an organic chemical synthesis intermediate.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
1-Boc-3-iodoazetidine is used as an organic chemical synthesis intermediate.

Notes
Light sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 254454-54-1
Molecular Formula C8H14INO2
Molecular Weight (g/mol) 283.109
MDL Number MFCD09752821
InChI Key XPDIKRMPZNLBAC-UHFFFAOYSA-N
Synonym 1-boc-3-iodoazetidine, 1-boc-3-iodo-azetidine, 1-boc-3-iodo azetidine, n-boc-3-iodoazetidine, 3-iodo-azetidine-1-carboxylic acid tert-butyl ester
PubChem CID 11000522
IUPAC Name tert-butyl 3-iodoazetidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CC(C1)I

Specifications

Quantity 5 g
Sensitivity Light sensitive
Formula Weight 283.11
Percent Purity 97%
Chemical Name or Material 1-Boc-3-iodoazetidine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement

H315-H319-H335

Causes skin irritation.

Causes serious eye irritation.

May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.