Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-BOC-4-chloropiperidine, 97%

Catalog No. AC435010010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC435010010 1 g
AC435010050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC435010010 Supplier Thermo Scientific Chemicals Supplier No. 435010010
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 154874-94-9 | C10H18ClNO2 | 219.71 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 154874-94-9
Molecular Formula C10H18ClNO2
Molecular Weight (g/mol) 219.71
MDL Number MFCD04115040
InChI Key NZZWXABIGMMKQL-UHFFFAOYSA-N
Synonym n-boc-4-chloro-piperidine, 1-boc-4-chloropiperidine, 1-boc-4-chloro-piperidine, 4-chloro-piperidine-1-carboxylic acid tert-butyl ester, n-boc-4-chloropiperidine, 1-piperidinecarboxylic acid, 4-chloro-, 1,1-dimethylethyl ester, chloropiperidine, 1-n-boc-4-chloropiperidine, ksc528i4h, 1-n-boc-4-chloro-piperidine
PubChem CID 11138624
IUPAC Name tert-butyl 4-chloropiperidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCC(Cl)CC1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 219.71
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 1-BOC-4-chloropiperidine
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/e

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.