Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Boc-6-cyanoindole-2-boronic acid, 96%

Catalog No. AAH52520MD
Click to view available options
Quantity:
250 mg
1 g

Catalog No. AAH52520MD

Supplier: Thermo Scientific Chemicals H52520MD

Only null left
Add to Cart

CAS: 913835-67-3 | C14H15BN2O4 | 286.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 913835-67-3
Molecular Formula C14H15BN2O4
Molecular Weight (g/mol) 286.09
MDL Number MFCD08436056
InChI Key CZEBTZMIQZVUSE-UHFFFAOYSA-N
Synonym 1-boc-6-cyanoindole-2-boronic acid, 1-tert-butoxycarbonyl-6-cyano-1h-indol-2-yl boronic acid, 1-tert-butoxycarbonyl-6-cyano-1h-indole-2-boronic acid, 6-cyano-1h-indole-2-boronic acid, n-boc protected, 1-tert-butoxycarbonyl-6-cyanoindol-2-ylboronic acid, acmc-209r9t, 6-cyano-1h-indol-2-ylboronic acid, n-boc protected, 1-tert-butoxycarbonyl-6-cyano-1h-indol-2-yl boronicacid, 1-tert-butoxycarbonyl-2-dihydroxyboryl-1h-indole-6-carbonitrile
PubChem CID 44119347
IUPAC Name [6-cyano-1-[(2-methylpropan-2-yl)oxycarbonyl]indol-2-yl]boronic acid
SMILES CC(C)(C)OC(=O)N1C(=CC2=CC=C(C=C12)C#N)B(O)O

Specifications

Melting Point 219°C to 221°C
Quantity 250 mg
Formula Weight 286.09
Percent Purity 96%
Chemical Name or Material 1-Boc-6-cyanoindole-2-boronic acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.