Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-Boc-azetidine-3-carboxylic acid | 142253-55-2 | MFCD01860897 | 98.3% | 201.22 g/mol | C9H15NO4 | 100 G
SDP

Supplier:  Medchemexpress LLC HY40141100G

Encompass_Preferred

1-Boc-azetidine-3-carboxylic acid is a Boc-protected azetidine derivative used as a non-cleavable ADC linker and as an alkyl chain-based PROTAC linker. It is supplied as a white to off-white solid with reported high purity, and it is provided in multiple laboratory-scale pack sizes with standard storage and solubility recommendations.

  • Non-cleavable ADC linker for antibody-drug conjugate synthesis.
  • Suitable as an alkyl chain-based PROTAC linker.
  • Appearance: white to off-white solid.
  • Purity approximately 98.3% as supplied.
  • Chemical formula C9H15NO4; molecular weight 201.22 g/mol.
  • High solubility in DMSO (reported 100 mg/mL) and formulation protocols for in vivo use.
  • Storage recommendations: powder at -20°C for long term, 4°C for shorter-term storage.

Catalog No. 50-002-04083


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.