Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Boc-indoline-7-carboxylic acid, 97%

Catalog No. AAH63883MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH63883MD 250 mg
AAH6388306 5 g
AAH6388303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH63883MD Supplier Thermo Scientific Chemicals Supplier No. H63883MD

CAS: 143262-20-8 | C14H17NO4 | 263.293 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 143262-20-8
Molecular Formula C14H17NO4
Molecular Weight (g/mol) 263.293
MDL Number MFCD04973983
InChI Key SUAMIYWLXFROHE-UHFFFAOYSA-N
Synonym n-boc-indoline-7-carboxylic acid, 1-tert-butoxycarbonyl indoline-7-carboxylic acid, 2,3-dihydro-indole-1,7-dicarboxylic acid 1-tert-butyl ester, 1-tert-butoxycarbonyl-2,3-dihydroindole-7-carboxylic acid, n-boc-indoline-7-carboxylicacid, acmc-1c2ys, 1-tert-butoxycarbonyl-7-indolinecarboxylic acid, 1h-indole-1,7-dicarboxylic acid,2,3-dihydro-,1-1,1-dimethylethyl ester, 11bs-2,6-bis 3,5-bis trifluoromethyl phenyl-4-hydroxy-dinaphtho 2,1-d:1 inverted exclamation marka,2 inverted exclamation marka-f-1,3,2-dioxaphosphepin 4-oxide
PubChem CID 14977845
IUPAC Name 1-[(2-methylpropan-2-yl)oxycarbonyl]-2,3-dihydroindole-7-carboxylic acid
SMILES CC(C)(C)OC(=O)N1CCC2=CC=CC(=C21)C(=O)O

Specifications

Melting Point 182°C (decomposition)
Color Yellow
Quantity 250 mg
UN Number UN3077
Sensitivity Light sensitive
Formula Weight 263.29
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 1-Boc-indoline-7-carboxylic acid
Hazard Category H317
Hazard Statement GHS H Statement
H317
May cause an allergic skin reaction.
Precautionary Statement P261-P272-P280g-P302+P352-P333+P313-P363-P501c
DOTInformation Transport Hazard Class: 9; Packaging Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.