Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-Bromo-2-(2-methoxyethoxy)ethane

Catalog No. 63984210G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
63-984-210G 10 g
63-984-250G 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 63-984-210G Supplier Sigma Aldrich Supplier No. 63984210G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 1-bromo-2-(2-methoxyethoxy)ethane can be used in the synthesis of 1,4-diketo-3,6-diphenylpyrrolo[3,4-c]pyrrole(dpp)derivatives, which are stable and efficient pigments for two-photon excited fluorescence microscopy
  • It can also be used in the synthesis of a squarine dye, which exhibits a selective fluorogenic response towards zinc cation

Specifications

CAS 54149-17-6
Density 1.3600 g/mL (at 25°C (literature))
Boiling Point 156°C (lit.), 37°C to 40°C (0.01 mmHg)
Molecular Formula C5H11BrO2
Refractive Index n20/D 1.455 (literature)
Linear Formula CH3(OCH2CH2)2 Br
MDL Number MFCD00000238
Quantity 10 g
Molecular Weight (g/mol) 183.04
Percent Purity 95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.