Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Bromo-2-(2-methoxyethoxy)ethane 98%, Thermo Scientific™

Catalog No. AC177660500
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC177660500 50 g
AC177660100 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC177660500 Supplier Thermo Scientific Chemicals Supplier No. 177660500

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54149-17-6
Molecular Formula C5H11BrO2
Molecular Weight (g/mol) 183.05
MDL Number MFCD00000238
InChI Key HUXJXNSHCKHFIL-UHFFFAOYSA-N
Synonym 1-bromo-2-2-methoxyethoxy ethane, 1-2-bromoethoxy-2-methoxyethane, 1-bromo-3,6-dioxaheptane, ethane, 1-2-bromoethoxy-2-methoxy, 2-2-methoxyethoxy ethyl bromide, 1-2-bromoethoxy-2-methoxy-ethane, 1-bromo-2-2-methoxy-ethoxy ethane, mpeg2-br, acmc-1alws, 7-bromo-2,5-dioxaheptane
PubChem CID 123532
IUPAC Name 1-bromo-2-(2-methoxyethoxy)ethane
SMILES COCCOCCBr

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless
Infrared Spectrum Authentic
Refractive Index 1.4550 to 1.4570 (20°C, 589nm)
Linear Formula CH3(OCH2CH2)2Br
Quantity 50 g
Formula Weight 183.04
Percent Purity ≥97.5%
Physical Form Liquid
Chemical Name or Material 1-Bromo-2-(2-methoxyethoxy)ethane, 98%
Show More Show Less
EINECSNumber 259-000-6
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.