Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Bromo-2-(2-methoxyethoxy)ethane (stabilized with Na2CO3) 90.0+%, TCI America™
SDP

Catalog No. B47365G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B47365G 5 g
B473625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B47365G Supplier TCI America Supplier No. B47365G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 54149-17-6
Molecular Formula C5H11BrO2
Molecular Weight (g/mol) 183.05
MDL Number MFCD00000238
InChI Key HUXJXNSHCKHFIL-UHFFFAOYSA-N
Synonym 1-bromo-2-2-methoxyethoxy ethane, 1-2-bromoethoxy-2-methoxyethane, 1-bromo-3,6-dioxaheptane, ethane, 1-2-bromoethoxy-2-methoxy, 2-2-methoxyethoxy ethyl bromide, 1-2-bromoethoxy-2-methoxy-ethane, 1-bromo-2-2-methoxy-ethoxy ethane, mpeg2-br, acmc-1alws, 7-bromo-2,5-dioxaheptane
PubChem CID 123532
IUPAC Name 1-bromo-2-(2-methoxyethoxy)ethane
SMILES COCCOCCBr

Specifications

Color Yellow
Boiling Point 156°C
Quantity 5 g
Formula Weight 183.05
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Bromo-2-(2-methoxyethoxy)ethane (stabilized with Na2CO3)
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.