Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Bromo-2,3,5-trifluorobenzene 98.0+%, TCI America™
SDP

Catalog No. B29701G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B29701G 1 g
B29705G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B29701G Supplier TCI America Supplier No. B29701G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 133739-70-5
Molecular Formula C6H2BrF3
Molecular Weight (g/mol) 210.98
MDL Number MFCD00012232
InChI Key XSMLLZPSNLQCQU-UHFFFAOYSA-N
PubChem CID 2736399
IUPAC Name 1-bromo-2,3,5-trifluorobenzene
SMILES FC1=CC(F)=C(F)C(Br)=C1

Specifications

Color Yellow
Quantity 1 g
UN Number 1993
Synonym 2,3,5-trifluorobromobenzene, benzene, 1-bromo-2,3,5-trifluoro, 1-bromo-2,3,5-trifluoro-benzene, pubchem2305, acmc-209bte, intermediates-zcf02035, 2,3,5-trifluorobromo benzene, 2,3,5-trifluorophenyl bromide, paragos 390204, benzene,1-bromo-2,3,5-trifluoro
Formula Weight 210.98
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Bromo-2,3,5-trifluorobenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.