Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Bromo-2,6-dichlorobenzene 98.0+%, TCI America™
SDP

Catalog No. B098225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B098225G 25 g
B09825G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B098225G Supplier TCI America Supplier No. B098225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 19393-92-1
Molecular Formula C6H3BrCl2
Molecular Weight (g/mol) 225.894
MDL Number MFCD00000574
InChI Key UWOIDOQAQPUVOH-UHFFFAOYSA-N
Synonym 1-bromo-2,6-dichlorobenzene, 2,6-dichlorobromobenzene, 1,3-dichloro-2-bromobenzene, benzene, 2-bromo-1,3-dichloro, 2,6-dichloro-1-bromobenzene, benzene, 1-bromo-2,6-dichloro, pubchem3610, acmc-209exg, 2,6-dichlorobromo benzene, 2,6-dichlorophenyl bromide
PubChem CID 29568
IUPAC Name 2-bromo-1,3-dichlorobenzene
SMILES C1=CC(=C(C(=C1)Cl)Br)Cl

Specifications

Melting Point 66°C
Color White-Yellow
Boiling Point 242°C
Quantity 25 g
Formula Weight 225.89
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Bromo-2,6-dichlorobenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.