Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium peroxide, ACS reagent

Catalog No. AC315765000
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC315765000 500 g Plastic bottle
AC315760250 25 g Plastic bottle
AC315761000 100 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC315765000 Supplier Thermo Scientific Chemicals Supplier No. 315765000
Only null left
Add to Cart

CAS: 1313-60-6 | Na2O2 | 77.97 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1313-60-6
Molecular Formula Na2O2
Molecular Weight (g/mol) 77.97
MDL Number MFCD00003497
InChI Key PFUVRDFDKPNGAV-UHFFFAOYSA-N
Synonym sodium peroxide, disodium peroxide, solozone, natriumsuperoxid, sodium dioxide, disodium dioxide, natrium peroxydatum, sodium oxide, per, flocool 180, sodium oxide
PubChem CID 14803
IUPAC Name disodium;peroxide
SMILES [O-][O-].[Na+].[Na+]

Specifications

Melting Point 460.0°C
Color Yellow
Physical Form Powder/Granules
Quantity 500 g
Linear Formula Na2O2
Fieser 06,548; 07,341; 11,492
Merck Index 15, 8786
Solubility Information Solubility in water: reacts violently. Other solubilities: forming naoh and h2o2,freely soluble
Formula Weight 77.97
Percent Purity ≥93.0%
Grade ACS Reagent
Packaging Plastic bottle
Specific Gravity 2.8
Density 2.8000g/mL
Chemical Name or Material Sodium peroxide
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
May cause fire or explosion; strong oxidizer.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
EINECSNumber 215-209-4

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.