Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Bromo-2-naphthoic Acid 98.0+%, TCI America™
SDP

Catalog No. B16175G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B16175G 5 g
B1617100MG 100 mg
B16171G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B16175G Supplier TCI America Supplier No. B16175G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 20717-79-7
Molecular Formula C11H7BrO2
Molecular Weight (g/mol) 251.079
MDL Number MFCD00021408
InChI Key VUVIRKAVBZITDO-UHFFFAOYSA-N
Synonym 1-bromo-2-naphthoic acid, 1-bromo-2-naphthalenecarboxylic acid, 2-naphthalenecarboxylic acid, 1-bromo, maybridge4_002377, 1-bromo-2-naphthoicacid, 1-bromo-2-napthoic acid, 1-bromo-2-naphtoic acid, acmc-1ck28, 2-?naphthalenecarboxyli?c acid, 2-naphthalenecarboxylicacid,1-bromo
PubChem CID 88665
IUPAC Name 1-bromonaphthalene-2-carboxylic acid
SMILES C1=CC=C2C(=C1)C=CC(=C2Br)C(=O)O

Specifications

Melting Point 191°C
Color White-Yellow
Quantity 5 g
Formula Weight 251.08
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Bromo-2-naphthoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.