Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Bromo-3,4,5-trifluorobenzene 98.0+%, TCI America™
SDP

Catalog No. B176525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B176525G 25 g
B17655G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B176525G Supplier TCI America Supplier No. B176525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 138526-69-9
Molecular Formula C6H2BrF3
Molecular Weight (g/mol) 210.981
MDL Number MFCD00070738
InChI Key HKJCELUUIFFSIN-UHFFFAOYSA-N
Synonym 1-bromo-3,4,5-trifluorobenzene, 3,4,5-trifluorobromobenzene, 1,2,3-trifluoro-5-bromobenzene, 5-bromo-1,2,3-trifluoro-benzene, benzene, 5-bromo-1,2,3-trifluoro, pubchem2304, acmc-1c02k, ksc174s3f, 3,4,5-trifluoro-bromobenzene, 3,4,5-trifluorophenyl bromide
PubChem CID 611409
IUPAC Name 5-bromo-1,2,3-trifluorobenzene
SMILES C1=C(C=C(C(=C1F)F)F)Br

Specifications

Color Yellow
Boiling Point 43°C
Quantity 25 g
UN Number 1993
Formula Weight 210.98
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Bromo-3,4,5-trifluorobenzene
EINECSNumber (3)-4279
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.