Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Bromo-3,5-di-tert-butylbenzene 98.0+%, TCI America™
SDP

Catalog No. B354725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B354725G 25 g
B35475G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B354725G Supplier TCI America Supplier No. B354725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 22385-77-9
Molecular Formula C14H21Br
Molecular Weight (g/mol) 269.226
MDL Number MFCD00796945
InChI Key BUOWTUULDKULFI-UHFFFAOYSA-N
Synonym 1-bromo-3,5-di-tert-butylbenzene, 3,5-di-tert-butylbromobenzene, 3,5-di-t-butylbromobenzene, 1-bromo-3,5-di-t-butylbenzene, 1-bromo-3,5-ditert-butyl benzene, benzene, 1-bromo-3,5-bis 1,1-dimethylethyl, 1,5-bis tert-butyl-3-bromobenzene, acmc-1cpbo, g00018-watson-int, 3,5-di-t-butyl-bromobenzene
PubChem CID 620136
IUPAC Name 1-bromo-3,5-ditert-butylbenzene
SMILES CC(C)(C)C1=CC(=CC(=C1)Br)C(C)(C)C

Specifications

Melting Point 65°C
Color White-Yellow
Boiling Point 158°C
Quantity 25 g
Formula Weight 269.23
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Bromo-3,5-di-tert-butylbenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.