Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Bromo-3,5-dimethoxybenzene 98.0+%, TCI America™
SDP

Catalog No. B384825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B384825G 25 g
B38485G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B384825G Supplier TCI America Supplier No. B384825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 20469-65-2
Molecular Formula C8H9BrO2
Molecular Weight (g/mol) 217.06
MDL Number MFCD06200685
InChI Key KRWRFIMBWRVMKE-UHFFFAOYSA-N
Synonym 3,5-dimethoxybromobenzene, benzene, 1-bromo-3,5-dimethoxy, 1,3-dimethoxy-5-bromobenzene, 1-bromo-3,5-dimethoxy-benzene, 1-bromo-3,5-dimethox, 3,5-dimethoxy-bromobenzene, bromo-3,5-dimethoxybenzene, 3,5-dimethoxy bromobenzene, acmc-1cs59, ksc495e7l
PubChem CID 639187
IUPAC Name 1-bromo-3,5-dimethoxybenzene
SMILES COC1=CC(OC)=CC(Br)=C1

Specifications

Melting Point 67°C
Color White-Yellow
Quantity 25 g
Formula Weight 217.06
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 1-Bromo-3,5-dimethoxybenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.