Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Bromo-3-chloro-2-methylpropane 98.0+%, TCI America™
SDP

Catalog No. B185925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B185925G 25 g
B1859500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B185925G Supplier TCI America Supplier No. B185925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6974-77-2
Molecular Formula C4H8BrCl
Molecular Weight (g/mol) 171.462
MDL Number MFCD00000955
InChI Key ZKDOQFPDSUOLGF-UHFFFAOYSA-N
PubChem CID 95566
IUPAC Name 1-bromo-3-chloro-2-methylpropane
SMILES CC(CCl)CBr

Specifications

Color Colorless
Boiling Point 154°C
Quantity 25 g
Formula Weight 171.46
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Bromo-3-chloro-2-methylpropane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.