Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Bromo-3-chloropropane, 99%

Catalog No. p-7081422
Change view
Click to view available options
Quantity:
100 g
250 g
1000 g
5000 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1139522 100 g
AAA1139530 250 g
AAA113950B 1000 g
AAA113950I 5000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AAA1139522 Supplier Thermo Scientific Chemicals Supplier No. A1139522
Only null left
Add to Cart

CAS: 109-70-6 | C3H6BrCl | 157.435 g/mol

1-Bromo-3-chloropropane is used in the preparation active pharmaceutical ingredient intermediate such as gemfibrozil and reproterol. It is also involved in the preparation of cardiovascular diseases and analgesic drugs materials. It is utilized as a phase separation reagent for the isolation of ribonucleic acid (RNA) in high quality. It is considered as a replacement of chloroform in nucleic acid separations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromo-3-chloropropane is used in the preparation active pharmaceutical ingredient intermediate such as gemfibrozil and reproterol. It is also involved in the preparation of cardiovascular diseases and analgesic drugs materials. It is utilized as a phase separation reagent for the isolation of ribonucleic acid (RNA) in high quality. It is considered as a replacement of chloroform in nucleic acid separations.

Solubility
Miscible with alcohols, chlorofom and ether. Immiscible with water.

Notes
Incompatible with strong oxidizing, reducing agents, amines, nitrides, azo/diazo compounds, alkali metals and epoxides.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-70-6
Molecular Formula C3H6BrCl
Molecular Weight (g/mol) 157.435
MDL Number MFCD00000998
InChI Key MFESCIUQSIBMSM-UHFFFAOYSA-N
Synonym trimethylene chlorobromide, 1-chloro-3-bromopropane, 3-bromopropyl chloride, 3-chloropropyl bromide, propane, 1-bromo-3-chloro, 3-chloro-1-bromopropane, 3-bromo-1-chloropropane, trimethylene bromide chloride, 1,3-chbp, 1,3-chlorbromopropane
PubChem CID 8006
IUPAC Name 1-bromo-3-chloropropane
SMILES C(CCl)CBr

Specifications

Melting Point -59°C
Density 1.598
Boiling Point 142°C to 145°C
Flash Point >140°C (284°F)
Refractive Index 1.4855
Quantity 100 g
UN Number UN2688
Beilstein 605278
Solubility Information Miscible with alcohols,chlorofom and ether. Immiscible with water.
Formula Weight 157.44
Percent Purity 99%
Chemical Name or Material 1-Bromo-3-chloropropane
Show More Show Less
Hazard Category H227-H302-H331-H335-H341
Hazard Statement GHS H Statement
H331-H302
Toxic if inhaled.
Harmful if swallowed.
Precautionary Statement P201-P202-P210-P235-P261-P264b-P270-P271-P281-P301+P312-P304+P340-P308+P313-P311-P330-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: 1-BROMO-3-CHLOROPROPANE
EINECSNumber 203-697-1
RTECSNumber TX4113000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.