Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Bromo-3-n-octylbenzene 97.0+%, TCI America™
SDP

Catalog No. B48171G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B48171G 1 g
B48175G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B48171G Supplier TCI America Supplier No. B48171G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 663949-34-6
Molecular Formula C14H21Br
Molecular Weight (g/mol) 269.226
InChI Key SVXNIBGYTOIBPP-UHFFFAOYSA-N
PubChem CID 67410013
IUPAC Name 1-bromo-3-octylbenzene
SMILES CCCCCCCCC1=CC(=CC=C1)Br

Specifications

Color Yellow
Boiling Point 120°C
Quantity 1 g
Formula Weight 269.23
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Bromo-3-n-octylbenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.