Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Bromo-4-nonylbenzene 95.0+%, TCI America™
SDP

Catalog No. B159625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B159625G 25 g
B15965G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B159625G Supplier TCI America Supplier No. B159625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 51554-94-0
Molecular Formula C15H23Br
Molecular Weight (g/mol) 283.25
MDL Number MFCD00061116
InChI Key LVCWOFRWEVUMNY-UHFFFAOYSA-N
Synonym 1-bromo-4-n-nonylbenzene, bromononylbenzene, 4-nonylbromobenzene, 4-n-nonylbromobenzene, 4-nonylphenyl bromide, acmc-209kuh, 4-bromo-1-nonylbenzene, 1-bromo-4-nonylbenzene
PubChem CID 12603095
IUPAC Name 1-bromo-4-nonylbenzene
SMILES CCCCCCCCCC1=CC=C(Br)C=C1

Specifications

Color Colorless
Boiling Point 190°C
Quantity 25 g
Formula Weight 283.25
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Bromo-4-nonylbenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.