Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Bromo-4-pentylbenzene 90.0+%, TCI America™
SDP

Catalog No. B160525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B160525G 25 g
B16055G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B160525G Supplier TCI America Supplier No. B160525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 51554-95-1
Molecular Formula C11H15Br
Molecular Weight (g/mol) 227.145
MDL Number MFCD00061113
InChI Key SGCJPYYTVBHQGE-UHFFFAOYSA-N
Synonym 4-pentylbromobenzene, 4-bromo-n-pentylbenzene, 1-bromo-4-n-pentylbenzene, 4-n-amylbromobenzene, 4-n-pentylbromobenzene, 1-bromo-4-pentyl-benzene, p-bromopentylbenzene, 4-bromopentylbenzene, 1-amyl-4-bromobenzene, benzene, 1-bromo-4-pentyl
PubChem CID 2735599
IUPAC Name 1-bromo-4-pentylbenzene
SMILES CCCCCC1=CC=C(C=C1)Br

Specifications

Color Yellow
Boiling Point 148°C
Quantity 25 g
Formula Weight 227.15
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Bromo-4-pentylbenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.