Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Bromo-4-tert-butylbenzene, 97%

Catalog No. AAA1592422
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1592422 100 g
AAA1592414 25 g
AAA1592436 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1592422 Supplier Thermo Scientific Chemicals Supplier No. A1592422
Add to Cart

CAS: 3972-65-4 | C10H13Br | 213.118 g/mol

1-Bromo-4-tert-butylbenzene was used in the synthesis of 4-tert-butyl-phenylboronic acid1, 1-deoxy analogs of CP-47,497 (n = 0 to 7) and 1-deoxy analogs of CP-55,940 (n = 0 to 7). It undergoes lithium-bromide exchange reactions with n-butyllithium and tert-butyllithium at 0°C in various solvents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromo-4-tert-butylbenzene was used in the synthesis of 4-tert-butyl-phenylboronic acid1, 1-deoxy analogs of CP-47,497 (n = 0 to 7) and 1-deoxy analogs of CP-55,940 (n = 0 to 7). It undergoes lithium-bromide exchange reactions with n-butyllithium and tert-butyllithium at 0°C in various solvents.

Solubility
Insoluble in water.

Notes
Store in cool dry place. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3972-65-4
Molecular Formula C10H13Br
Molecular Weight (g/mol) 213.118
MDL Number MFCD00000108
InChI Key XHCAGOVGSDHHNP-UHFFFAOYSA-N
Synonym 4-tert-butylbromobenzene, 1-bromo-4-tert-butyl benzene, butylbromobenzene, p-bromo-tert-butylbenzene, 1-bromo-4-t-butylbenzene, 1-bromo-4-1,1-dimethylethyl benzene, 4-tert-butyl-1-bromobenzene, p-bromo-t-butylbenzene, 1-bromo-p-t-butylbenzene, 4-bromo-tert-butylbenzene
PubChem CID 77595
IUPAC Name 1-bromo-4-tert-butylbenzene
SMILES CC(C)(C)C1=CC=C(C=C1)Br

Specifications

Melting Point 15°C to 16°C
Density 1.229
Boiling Point 231°C to 232°C
Flash Point 97°C (206°F)
Refractive Index 1.533
Quantity 100 g
Beilstein 1859117
Solubility Information Insoluble in water.
Formula Weight 213.12
Percent Purity 97%
Chemical Name or Material 1-Bromo-4-tert-butylbenzene
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 223-599-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.