Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Bromo-4-tert-butylbenzene 96.0+%, TCI America™
SDP

Catalog No. B1727100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B1727100G 100 g
B172725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B1727100G Supplier TCI America Supplier No. B1727100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3972-65-4
Molecular Formula C10H13Br
Molecular Weight (g/mol) 213.118
MDL Number MFCD00000108
InChI Key XHCAGOVGSDHHNP-UHFFFAOYSA-N
Synonym 4-tert-butylbromobenzene, 1-bromo-4-tert-butyl benzene, butylbromobenzene, p-bromo-tert-butylbenzene, 1-bromo-4-t-butylbenzene, 1-bromo-4-1,1-dimethylethyl benzene, 4-tert-butyl-1-bromobenzene, p-bromo-t-butylbenzene, 1-bromo-p-t-butylbenzene, 4-bromo-tert-butylbenzene
PubChem CID 77595
IUPAC Name 1-bromo-4-tert-butylbenzene
SMILES CC(C)(C)C1=CC=C(C=C1)Br

Specifications

Color Yellow
Boiling Point 228°C
Quantity 100 g
Formula Weight 213.12
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Bromo-4-tert-butylbenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.