Learn More
Sigma Organic Chemistry 1-Bromobutane | 1KG | 109-65-9 | MFCD00000260
Supplier: Sigma Organic Chemistry B594971KG

1-Bromobutane, 1KG
About This Item:
Synonym(s): Butyl bromide
Linear Formula: CH3(CH2)3Br
Molecular Weight: 137.02
CAS Number: 109-65-9
MDL Number: MFCD00000260
UNSPSC Code: 12352100
Purity: 0.99
Boiling Point: 100-104 C (lit.)
Melting Point: -112 C (lit.)
Density: 1.276 g/mL at 25 C (lit.)
Refractive Index: n20/D 1.439 (lit.)
Storage: room temp
EINECS Number: 203-691-9
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.