Learn More
1-Bromoheptane, 98+%
Description
1-bromoheptane is an important precursor material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
1-bromoheptane is an important precursor material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.
Solubility
Immiscible with water.
Notes
Flammable and incompatible with oxidizing agents, strong bases.
Specifications
Specifications
| Melting Point | -58°C |
| Density | 1.139 |
| Boiling Point | 179°C to 180°C |
| Flash Point | 60°C (140°F) |
| Odor | Recognizable |
| Refractive Index | 1.4499 |
| Quantity | 5000 g |
| UN Number | UN1993 |
| Beilstein | 1697160 |
| Solubility Information | Immiscible with water. |
| Show More |
Safety and Handling
| Hazard Category | H226-H315-H319-H332-H335 |
| Hazard Statement | GHS H Statement H226-H332-H315-H319-H335 Flammable liquid and vapour. Harmful if inhaled. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S. |
| EINECSNumber | 211-068-8 |
| RTECSNumber | MI8100000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.