Learn More
Sigma Organic Chemistry 1-Bromomethyl-2 2-diflu | 1G | 77613-65-1 | MFCD06797749
Supplier: Sigma Organic Chemistry MNO0001781G

1-Bromomethyl-2 2-diflu , 1G
About this item:
CAS #: 77613-65-1
MDL #: MFCD06797749
Chemical Formula: C4H5BrF2
Molecular Weight: 170.98
UNSPSC Code: 12352200
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.