Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(bromomethyl)-2-phenoxybenzene, 90%, Thermo Scientific™

Catalog No. CC49908CB
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC49908CB 250 mg
CC49908DA 1 g
CC49908DE 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC49908CB Supplier Thermo Scientific Chemicals Supplier No. CC49908CB
Add to Cart

Chemical Identifiers

CAS 82657-72-5
Molecular Formula C13H11BrO
Molecular Weight (g/mol) 263.134
InChI Key YQRIQBOWLXRKKG-UHFFFAOYSA-N
Synonym 1-bromomethyl-2-phenoxybenzene, 2-phenoxybenzyl bromide, benzene, bromomethylphenoxy, acmc-20eqyy, 2-bromomethylphenyl phenyl ether, 1-bromomethyl-2-phenoxy-benzene, 2-bromomethyl-1-phenoxybenzene, 1-2-bromomethyl phenoxy benzene, 1-bromomethyl-2-phenyloxy benzene, benzene, 1-bromomethyl-2-phenoxy
PubChem CID 22675469
IUPAC Name 1-(bromomethyl)-2-phenoxybenzene
SMILES C1=CC=C(C=C1)OC2=CC=CC=C2CBr

Specifications

Assay Percent Range 97%
Quantity 250 mg
Formula Weight 263.13
Percent Purity 97%
Chemical Name or Material 1-(Bromomethyl)-2-phenoxybenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.