Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(Bromomethyl)naphthalene, 98%, Thermo Scientific Chemicals

Catalog No. AAH6411622
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6411622 100 g
AAH6411614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6411622 Supplier Thermo Scientific Chemicals Supplier No. H6411622

It is employed as a intermediate for organic synthesis and pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a intermediate for organic synthesis and pharmaceutical.

Solubility
Slightly soluble in water.

Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3163-27-7
Molecular Formula C11H9Br
Molecular Weight (g/mol) 221.097
MDL Number MFCD00010804
InChI Key RZJGKPNCYQZFGR-UHFFFAOYSA-N
Synonym 1-bromomethyl naphthalene, 1-bromomethyl-naphthalene, naphthalene, 1-bromomethyl, 1-bromomethy naphthalene, naphtylmethylbromide, bromomethylnaphthalene, naphthylmethyl bromide, pubchem9389
PubChem CID 137844
IUPAC Name 1-(bromomethyl)naphthalene
SMILES C1=CC=C2C(=C1)C=CC=C2CBr

Specifications

Color White
Density 1.44 g/mL
Boiling Point 213°C (100 mmHg)
Refractive Index 1.66
Quantity 100 g
UN Number UN3261
Beilstein 742417
Solubility Information Slightly soluble in water.
Formula Weight 221.1
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 1-(Bromomethyl)naphthalene
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packaging Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 801-817-2
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.