Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Bromonaphthalene, 97%

Catalog No. aaa1412218
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
1000 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1412218 50 g
AAA1412209 10 g
AAA1412230 250 g
AAA141220B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AAA1412218 Supplier Thermo Scientific Chemicals Supplier No. A1412218
Only null left
Add to Cart

CAS: 90-11-9 | C10H7Br | 207.07 g/mol

1-Bromonaphthalene is used in organic synthesis and refractometry of fats. It is used in the preparation of N-aryl imidazoles and diaryl ethers. It is used to determine the refractive index of crystals and the water content of alcohols. It is utilized in the preparation of glutathione peroxidase-like antioxidant activity of diaryl diselenides. Further, it serves as a solvent for the exfoliation and dispersion of hexabenzocoronene. In addition to this, it is used as a refrigerant and a solvent molecular weight substance.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromonaphthalene is used in organic synthesis and refractometry of fats. It is used in the preparation of N-aryl imidazoles and diaryl ethers. It is used to determine the refractive index of crystals and the water content of alcohols. It is utilized in the preparation of glutathione peroxidase-like antioxidant activity of diaryl diselenides. Further, it serves as a solvent for the exfoliation and dispersion of hexabenzocoronene. In addition to this, it is used as a refrigerant and a solvent molecular weight substance.

Solubility
Miscible with chloroform, ethanol, ether, benzene and ethyl acetate. Slightly miscible with water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90-11-9
Molecular Formula C10H7Br
Molecular Weight (g/mol) 207.07
MDL Number MFCD00003868
InChI Key DLKQHBOKULLWDQ-UHFFFAOYSA-N
Synonym naphthalene, 1-bromo, 1-naphthyl bromide, alpha-bromonaphthalene, bromonaphthalene, alpha-naphthyl bromide, .alpha.-bromonaphthalene, 1-bromo-naphthalene, .alpha.-naphthyl bromide, naphthalene, bromo, bromonapthalene
PubChem CID 7001
IUPAC Name 1-bromonaphthalene
SMILES C1=CC=C2C(=C1)C=CC=C2Br

Specifications

Melting Point °C to 2°C to 0°C
Density 1.485
Boiling Point 279°C to 281°C
Flash Point >110°C (230°F)
Assay Percent Range 97%
Refractive Index 1.657
Quantity 50 g
Beilstein 1906414
Merck Index 14,1425
Solubility Information Miscible with chloroform,ethanol,ether,benzene and ethyl acetate. Slightly miscible with water.
Formula Weight 207.08
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1-Bromonaphthalene
Show More Show Less
Hazard Category H302
Hazard Statement GHS H Statement
H302-H319
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 201-965-2
RTECSNumber QJ1545000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.