Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Bromononane 98.0+%, TCI America™
SDP

Catalog No. B0933100ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0933100ML 100 mL
B093325ML 25 mL
B0933500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B0933100ML Supplier TCI America Supplier No. B0933100ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 693-58-3
Molecular Formula C9H19Br
Molecular Weight (g/mol) 207.155
MDL Number MFCD00000278
InChI Key AYMUQTNXKPEMLM-UHFFFAOYSA-N
Synonym n-nonyl bromide, nonyl bromide, nonane, 1-bromo, 1-nonyl bromide, 1-n-nonyl bromide, n-nonyl-1-bromide, bromononane, nonylbromide, 1-bromo-n-nonane, labotest-bb ltbb001163
PubChem CID 12742
IUPAC Name 1-bromononane
SMILES CCCCCCCCCBr

Specifications

Color Yellow
Boiling Point 201°C
Quantity 100 mL
Formula Weight 207.16
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Bromononane
EINECSNumber (2)-0067
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.