Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Bromopentadecane 98.0+%, TCI America™
SDP

Catalog No. B0936250ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0936250ML 250 mL
B093625ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B0936250ML Supplier TCI America Supplier No. B0936250ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 629-72-1
Molecular Formula C15H31Br
Molecular Weight (g/mol) 291.32
MDL Number MFCD00000229
InChI Key JKOTZBXSNOGCIF-UHFFFAOYSA-N
PubChem CID 12394
IUPAC Name 1-bromopentadecane
SMILES CCCCCCCCCCCCCCCBr

Specifications

Color Yellow
Boiling Point 158°C
Quantity 250 mL
Synonym pentadecyl bromide, pentadecane, 1-bromo, 1-bromo pentadecane, n-pentadecyl bromide, pentadecane,1-bromo, 1-bromopentadecane, acmc-209nb8, dsstox_cid_29296, dsstox_rid_83414, dsstox_gsid_49339
Formula Weight 291.32
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Bromopentadecane
EINECSNumber (2)-0067
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.