Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Bromoperfluoro-4,7,10,13-tetramethyl-3,6,9,12,15-pentaoxaheptadecane 94.0+%, TCI America™
SDP

Catalog No. B41671G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B41671G 1 g
B41675G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B41671G Supplier TCI America Supplier No. B41671G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1482416-52-3
Molecular Formula C16BrF33O5
Molecular Weight (g/mol) 979.02
MDL Number MFCD27918480
InChI Key VAXWUAHPDZRLAP-UHFFFAOYNA-N
Synonym 1-Bromoperfluoro-4,7,10,13-tetramethyl-3,6,9,12,15-pentaoxaheptadecane
PubChem CID 91972086
IUPAC Name 1-bromo-1,1,2,2,4,5,5,7,8,8,10,11,11,13,14,14,16,16,17,17,17-henicosafluoro-4,7,10,13-tetrakis(trifluoromethyl)-3,6,9,12,15-pentaoxaheptadecane
SMILES FC(F)(F)C(F)(F)OC(F)(F)C(F)(OC(F)(F)C(F)(OC(F)(F)C(F)(OC(F)(F)C(F)(OC(F)(F)C(F)(F)Br)C(F)(F)F)C(F)(F)F)C(F)(F)F)C(F)(F)F

Specifications

Color Colorless
Quantity 1 g
Formula Weight 979.02
Percent Purity ≥94.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Bromo-1,1,2,2,4,5,5,7,8,8,10,11,11,13,14,14,16,16,17,17,17-henicosafluoro-4,7,10,13-tetrakis(trifl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.