Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Bromotridecane 98.0+%, TCI America™
SDP

Catalog No. B093525ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B093525ML 25 mL
B09355ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B093525ML Supplier TCI America Supplier No. B093525ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 765-09-3
Molecular Formula C13H27Br
Molecular Weight (g/mol) 263.26
MDL Number MFCD00000227
InChI Key BFDNZQUBFCYTIC-UHFFFAOYSA-N
Synonym Tridecyl Bromide
PubChem CID 13000
IUPAC Name 1-bromotridecane
SMILES CCCCCCCCCCCCCBr

Specifications

Color Yellow
Boiling Point 150°C
Quantity 25 mL
Formula Weight 263.26
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Bromotridecane
EINECSNumber (2)-0067
RTECSNumber YD3100000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.