Learn More
Description
- We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories: Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications. Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
Specifications
Specifications
| Melting Point | ≤-90°C |
| Color | Colorless |
| Appearance | Liquid |
| Density | 0.81g/mL at 25°C |
| Quantity | 2.5 L |
| Explosion Limit | 11.2% |
| Ignition Point | 649°C |
| Packaging | Glass Bottle |
| Linear Formula | CH3(CH2)3OH |
| UN Number | UN1120 |
| Show More |
Safety and Handling
| Hazard Statement | P210-P280-P301 + P312 + P330-P304 + P340 + P312-P305 + P351 + P338 + P310-P403 + P235 |
| DOTInformation | class 3 - PG 3 - Butanols |
| RTECSNumber | 200-751-6 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.