Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Butanol, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., ≥99.5% (GC), Solstice

Catalog No. 60046185 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
30 kg
4 x 2.5 L
6 x 1 L
Packaging:
Not available
Poly Bottle
Stainless Steel Can
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-046-185 30 kg Stainless Steel Can
60-046-183 1 L Poly Bottle
60-046-184 2.5 L Poly Bottle
60-046-187 6 x 1 L Poly Bottle
60-046-186 4 x 2.5 L Not available
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 60-046-185 Supplier Solstice Supplier No. 3306530KG
Add to Cart
Add to Cart

Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur.,≥99.5% (GC)

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71-36-3
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.123
MDL Number MFCD00002964
InChI Key LRHPLDYGYMQRHN-UHFFFAOYSA-N
Synonym 1-butanol, butanol, n-butanol, butyl alcohol, n-butyl alcohol, 1-hydroxybutane, propylcarbinol, butyl hydroxide, propylmethanol, methylolpropane
PubChem CID 263
ChEBI CHEBI:28885
IUPAC Name butan-1-ol
SMILES CCCCO

Specifications

Color Colorless
Density 0.81g/mL at 25°C
Quantity 30 kg
Explosion Limit 0.112
Flash Point 35°C (95°F)
Ignition Point 649°F
Packaging Stainless Steel Can
Refractive Index n20/D 1.399
Linear Formula CH3(CH2)3OH
UN Number UN1120
Beilstein 969148
Solubility Information 66g/L in water at 20°C, pH 7
Formula Weight 74.12g/mol
Percent Purity ≥99.5% (GC)
Grade ACS/puriss. p.a.
Chemical Name or Material 1-Butanol, Reag. Ph. Eur. reag. ISO
Show More Show Less
Hazard Category Danger
Hazard Statement P210-P280-P301 + P312 + P330-P304 + P340 + P312-P305 + P351 + P338 + P310-P403 + P235
Precautionary Statement H226-H302-H315-H318-H335-H336
DOTInformation Class 3 - PG 3 - Butanols
EINECSNumber 200-751-6
RTECSNumber EO1400000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.