Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Butyl-1-methylpyrrolidinium Bromide 97.0+%, TCI America™
SDP

Catalog No. B342725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B342725G 25 g
B34275G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B342725G Supplier TCI America Supplier No. B342725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 93457-69-3
Molecular Formula C9H20BrN
Molecular Weight (g/mol) 222.17
MDL Number MFCD07784439
InChI Key LCZRPQGSMFXSTC-UHFFFAOYSA-M
PubChem CID 11009532
IUPAC Name 1-butyl-1-methylpyrrolidin-1-ium;bromide
SMILES CCCC[N+]1(CCCC1)C.[Br-]

Specifications

Melting Point 214°C
Color White-Yellow
Quantity 25 g
Formula Weight 222.17
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Butyl-1-methylpyrrolidinium Bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.