Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Butyl-3-methylimidazolium Dicyanamide 96.0+%, TCI America™
SDP

Catalog No. B544725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B544725G 25 g
B54475G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B544725G Supplier TCI America Supplier No. B544725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 448245-52-1
Molecular Formula C10H15N5
Molecular Weight (g/mol) 205.265
MDL Number MFCD07784457
InChI Key ICIVTHOGIQHZRY-UHFFFAOYSA-N
Synonym 1-butyl-3-methylimidazolium dicyanamide, 1-butyl-3-methylimidazolium dicyamide, 1-butyl-3-methylimidazolium; dicyanoazanide, bmim dca, dsstox_cid_27886, dsstox_rid_82639, dsstox_gsid_47910, c4mim; dicyanoazanide, 1-butyl-3-methyl-1h-imidazol-3-ium cyanoimino methylidene azanide, 1-butyl-3-methylimidazolium; cyanoimino methylidene azanide
PubChem CID 11171745
IUPAC Name 1-butyl-3-methylimidazol-3-ium;cyanoiminomethylideneazanide
SMILES CCCCN1C=C[N+](=C1)C.C(=[N-])=NC#N

Specifications

Melting Point -6°C
Quantity 25 g
Formula Weight 205.27
Percent Purity ≥96.0% (HPLC,N)
Physical Form Liquid
Chemical Name or Material 1-Butyl-3-methylimidazolium Dicyanamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.