Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Butyl-3-methylimidazolium Methanesulfonate 98.0+%, TCI America™
SDP

Catalog No. B323125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B323125G 25 g
B32315G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B323125G Supplier TCI America Supplier No. B323125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 342789-81-5
Molecular Formula C9H18N2O3S
Molecular Weight (g/mol) 234.314
MDL Number MFCD06798173
InChI Key PUHVBRXUKOGSBC-UHFFFAOYSA-M
Synonym 1-butyl-3-methylimidazolium methanesulfonate, 1-butyl-3-methylimidazolium mesylate, acmc-20aowi, c4mim mesylate, basionics™ st 78, dsstox_cid_27888, dsstox_rid_82641, dsstox_gsid_47912, 1-butyl-3-methylimidazolium methylsulfonate, 1-n-butyl-3-methylimidazolium methanesulfonate
PubChem CID 11492381
IUPAC Name 1-butyl-3-methylimidazol-3-ium;methanesulfonate
SMILES CCCCN1C=C[N+](=C1)C.CS(=O)(=O)[O-]

Specifications

Melting Point 76°C
Color White-Yellow
Quantity 25 g
UN Number 3261
Formula Weight 234.31
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 1-Butyl-3-methylimidazolium Methanesulfonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.