Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Butyl-3-methylimidazolium Methyl Sulfate 98.0+%, TCI America™
SDP

Catalog No. B552725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B552725G 25 g
B55275G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B552725G Supplier TCI America Supplier No. B552725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 401788-98-5
Molecular Formula C9H18N2O4S
Molecular Weight (g/mol) 250.31
MDL Number MFCD03095437
InChI Key MEMNKNZDROKJHP-UHFFFAOYSA-M
Synonym 1-butyl-3-methylimidazolium methyl sulfate, 1-butyl-3-methylimidazolium methylsulfate, bmimmeso4, 1-n-butyl-3-methylimidazolium methylsulfate, 1-butyl-3-methylimidazolium methyl sulfate 1-, acmc-20aowh, basionics™ lq 02, 1-butyl-3-methylimidazolium methylsul-fate, 1-butyl-3-methylimidazol-3-ium,methyl sulfate, 1-butyl-3-methyl-imidazol-3-ium; methyl sulfate
PubChem CID 12095228
IUPAC Name 1-butyl-3-methyl-1H-imidazol-3-ium methyl sulfate
SMILES COS([O-])(=O)=O.CCCCN1C=C[N+](C)=C1

Specifications

Melting Point -4°C
Quantity 25 g
UN Number 3265
Formula Weight 250.31
Percent Purity ≥98.0% (HPLC)
Physical Form Liquid
Chemical Name or Material 1-Butyl-3-methylimidazolium Methyl Sulfate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.